2-Chlorothiophene-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A174695-1G A174695-250MG A174695-25G A174695-5G
76784-372EA 26.53 USD
76784-372 76784-374 76784-376 76784-378
2-Chlorothiophene-3-carboxylic acid 98%
2-Chlorothiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃ClO₂S
MW: 162.5941 g/mol
MDL Number: MFCD00098423
CAS Number: 53935-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR