Methyl-7-bromo-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

A174472-250MG A174472-5G A174472-1G A174472-25G
76784-356EA 16 USD
76784-356 76784-358 77871-872 77871-873
Methyl-7-bromo-1H-indole-2-carboxylate 98%
Methyl-7-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MDL Number: MFCD11111832
CAS Number: 1158503-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR