tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-(hydroxymethyl)-1H-indole, N-BOC protected, tert-Butyl 5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate

A174193-100MG A174193-1G
76784-346EA 9.55 USD
76784-346 76784-348
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₆BrNO₃
MW: 326.1857 g/mol
MDL Number: MFCD05864710
CAS Number: 905710-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR