Tetrabutylammonium chloride xhydrate 98%

Supplier: Ambeed
Warning

A173950-500G A173950-25G A173950-1000G A173950-100G
76784-330EA 55 USD
76784-330 77082-476 77667-420 77667-422
Tetrabutylammonium chloride xhydrate 98%
Tetrabutylammonium chloride xhydrate

Precaución: for Research Use Only

Formula: C₁₆H₃₆ClN·xH₂O
MDL Number: MFCD00011635
CAS Number: 37451-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR