trans-2-Chloromethylvinylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-chloro propenyl)dihydroxy borane, (3-Chloroprop-1-en-1-yl)boronic acid

A173467-100MG A173467-1G A173467-250MG A173467-5G
76784-306EA 23.34 USD
76784-306 76784-308 76784-310 76784-312
trans-2-Chloromethylvinylboronic acid 98%
trans-2-Chloromethylvinylboronic acid

Precaución: for Research Use Only

Formula: C₃H₆BClO₂
MW: 120.339 g/mol
MDL Number: MFCD01074546
CAS Number: 215951-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR