Ethyl-4-bromo-1H-indole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-bromo-1H-indole-2-carboxylate

A172228-100MG A172228-10G A172228-250MG A172228-5G A172228-1G
76784-284EA 19.1 USD
76784-284 76784-286 76784-288 76784-290 77667-404
Ethyl-4-bromo-1H-indole-2-carboxylate 95%
Ethyl-4-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.1066 g/mol
MDL Number: MFCD12174997
CAS Number: 103858-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR