Methyl-5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A172172-10G A172172-1G A172172-250MG A172172-5G
76784-274EA 12.74 USD
76784-274 76784-276 76784-278 76784-280 76784-282
Methyl-5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate 95%
Methyl-5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₄S
MDL Number: MFCD07782035
CAS Number: 914349-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR