3-Amino-6-chloropyrazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A172057-10G A172057-250MG A172057-25G
76784-264EA 181.42 USD
76784-264 76784-266 76784-268
3-Amino-6-chloropyrazine-2-carboxylic acid 95%
3-Amino-6-chloropyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C5H4ClN3O2
MW: 173.5600 g/mol
MDL Number: MFCD10697761
CAS Number: 2727-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR