N-Boc-2-bromo-D-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-Phe(2-Br)-OH
A171994-1G
A171994-250MG
76784-260EA
18
USD
76784-260
76784-262
N-Boc-2-bromo-D-phenylalanine 97%
N-Boc-2-bromo-D-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BrNO₄ MW: 344.205 g/mol |
MDL Number:
MFCD01317703 CAS Number: 261360-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...