3-Chloro-5-nitropyridin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-chloro-5-nitropyridine

A171699-100MG A171699-1G A171699-250MG A171699-5G
76784-248EA 27.59 USD
76784-248 76784-250 76784-252 76784-254
3-Chloro-5-nitropyridin-4-amine 97%
3-Chloro-5-nitropyridin-4-amine

Precaución: for Research Use Only

Formula: C5H4ClN3O2
MW: 173.5572 g/mol
MDL Number: MFCD14706168
CAS Number: 89284-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR