Z-Val-Ala-OH 98%

Supplier: Ambeed
Warning

A171269-1G A171269-250MG
76784-234EA 20.16 USD
76784-234 76784-236
Z-Val-Ala-OH 98%
Z-Val-Ala-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₅
MDL Number: MFCD00020395
CAS Number: 24787-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR