Trioctylamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tri-n-octylamine
A170756-100G
A170756-25G
A170756-1000G
A170756-500G
76784-224EA
12.74
USD
76784-224
77301-072
77567-506
77593-062
Trioctylamine 97%
Trioctylamine
Precaución: For research use only.
Formula:
C₂₄H₅₁N MW: 353.68 g/mol |
MDL Number:
MFCD00009560 CAS Number: 1116-76-3 UN: 3082 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...