(3-(Dimethylamino)propyl)triphenylphosphonium bromide hydrobromide 97%
Supplier: Ambeed
|
Warning
|
A170210-100G
A170210-10G
A170210-500G
A170210-5G
76784-182EA
38.2
USD
76784-182
76784-184
76784-186
76784-188
(3-(Dimethylamino)propyl)triphenylphosphonium bromide hydrobromide 97%
(3-(Dimethylamino)propyl)triphenylphosphonium bromide hydrobromide
Precaución: for Research Use Only
Formula:
C₂₃H₂₈Br₂NP MW: 509.25 g/mol |
MDL Number:
MFCD00077740 CAS Number: 27710-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...