5-Chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A170192-100MG A170192-1G A170192-250MG
76784-176EA 43.5 USD
76784-176 76784-178 76784-180
5-Chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 98%
5-Chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD09878688
CAS Number: 800401-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR