Boc-2-methyl-L-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Phe(2-Me)-OH
A169838-1G
A169838-250MG
A169838-5G
A169838-25G
76784-164EA
93.36
USD
76784-164
76784-166
76784-168
77667-370
77871-851
Boc-2-methyl-L-phenylalanine 97%
Boc-2-methyl-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₅H₂₁NO₄ MW: 279.34 g/mol |
MDL Number:
MFCD00671390 CAS Number: 114873-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...