(2-Chlorothiazol-5-yl)methylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2-Chlorothiazol-5-yl)methanamine hydrochloride

A169747-250MG A169747-5G
76784-156EA 12.74 USD
76784-156 76784-158
(2-Chlorothiazol-5-yl)methylamine hydrochloride 95%
(2-Chlorothiazol-5-yl)methylamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₆Cl₂N₂S
MW: 185.08 g/mol
MDL Number: MFCD09039253
CAS Number: 153471-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR