Tetrabutylammonium iodide 97%

Supplier: Ambeed
Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

A169033-1000G A169033-100G A169033-25G A169033-500G A169033-5G
76784-146EA 132.62 USD
76784-146 76784-148 76784-150 76784-152 76784-154
Tetrabutylammonium iodide 97%
Tetrabutylammonium iodide

Precaución: for Research Use Only

Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
MDL Number: MFCD00011636
CAS Number: 311-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR