Boc-3,4-dichloro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(3,4-DiCl)-OH

A168015-1G A168015-250MG A168015-25G A168015-5G
76784-122EA 48 USD
76784-122 76784-124 77667-346 77667-348
Boc-3,4-dichloro-L-phenylalanine 97%
Boc-3,4-dichloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.2 g/mol
MDL Number: MFCD00273436
CAS Number: 80741-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR