tert-Butyl 5-(aminomethyl)indoline-1-carboxylate 97%

Supplier: Ambeed
Warning

A167819-100MG A167819-1G A167819-250MG
76784-118EA 29 USD
76784-118 76784-120 77667-340
tert-Butyl 5-(aminomethyl)indoline-1-carboxylate 97%
tert-Butyl 5-(aminomethyl)indoline-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.32 g/mol
MDL Number: MFCD08234590
CAS Number: 885270-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR