Tirofiban 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Butylsulphonamido)-3-(4-(4-(piperidin-4-yl)butoxy)phenyl)propanoic acid, MK-383, L-7462

A167431-10MG A167431-1MG A167431-5MG A167431-1G A167431-250MG A167431-100MG
76784-104EA 62.6 USD
76784-104 76784-106 76784-108 77600-342 77600-344 77667-336
Tirofiban 98%
Tirofiban

Precaución: for Research Use Only

Formula: C₂₂H₃₆N₂O₅S
MW: 440.60 g/mol
MDL Number: MFCD05237246
CAS Number: 144494-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR