(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane) 97%
Supplier: Ambeed
|
Warning
|
A166935-1G
A166935-5G
76784-088EA
30.77
USD
76784-088
76784-090
(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane) 97%
(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane)
Precaución: for Research Use Only
Formula:
C₁₇H₃₂O MW: 252.44 g/mol |
MDL Number:
MFCD22421614 CAS Number: 95756-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...