5-Bromo-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A166494-100MG A166494-1G A166494-250MG
76784-074EA 77.45 USD
76784-074 76784-076 76784-078
5-Bromo-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid 98%
5-Bromo-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD11040726
CAS Number: 916325-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR