2-Amino-3-bromo-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-bromo-5-(trifluoromethyl)pyridine, 3-Bromo-5-(trifluoromethyl)pyridin-2-amine, 3-Bromo-5-(trifluoromethyl)-2-pyridinylamine, 2-Amino-3-bromo-5-trifluoromethylpyridine

A166451-1G A166451-25G A166451-5G
76784-068EA 8 USD
76784-068 76784-070 76784-072
2-Amino-3-bromo-5-(trifluoromethyl)pyridine 98%
2-Amino-3-bromo-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂
MW: 241.01 g/mol
MDL Number: MFCD07375382
CAS Number: 79456-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR