Methyl-2-chlorooxazole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chlorooxazole-4-carboxylate

A164977-10G A164977-1G A164977-250MG A164977-5G
76784-032EA 307.67 USD
76784-032 76784-034 76784-036 76784-038
Methyl-2-chlorooxazole-4-carboxylate 98%
Methyl-2-chlorooxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₄ClNO₃
MW: 161.55 g/mol
MDL Number: MFCD09265505
CAS Number: 934236-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR