(R)-1-tert-Butyl-2-methyl-5-oxopyrrolidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-tert-Butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate

A164851-100G A164851-10G A164851-1G A164851-25G A164851-5G
76784-018EA 132 USD
76784-018 76784-020 76784-022 76784-024 76784-026
(R)-1-tert-Butyl-2-methyl-5-oxopyrrolidine-1,2-dicarboxylate 97%
(R)-1-tert-Butyl-2-methyl-5-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₅
MDL Number: MFCD07783728
CAS Number: 128811-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR