Veliparib 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(2-Methylpyrrolidin-2-yl)-1H-benzo[d]imidazole-7-carboxamide

A164131-10MG A164131-1MG A164131-25MG A164131-5MG A164131-1G A164131-100MG A164131-50MG A164131-5G
76783-998EA 12.74 USD
76783-998 76784-000 76784-002 76784-004 77567-394 77667-296 77667-298 77667-300
Veliparib 98%
Veliparib

Precaución: For research use only.

Formula: C₁₃H₁₆N₄O
MW: 244.29 g/mol
MDL Number: MFCD16661059
CAS Number: 912444-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR