3-(Methylsulfonyl)aniline 95%
Supplier: Ambeed
|
Warning
|
A163578-100MG
A163578-10G
A163578-1G
A163578-250MG
A163578-25G
A163578-5G
76783-972EA
9.55
USD
76783-972
76783-974
76783-976
76783-978
76783-980
76783-982
3-(Methylsulfonyl)aniline 95%
3-(Methylsulphonyl)aniline
Precaución: for Research Use Only
Formula:
C₇H₉NO₂S MW: 171.2200 g/mol |
MDL Number:
MFCD03764428 CAS Number: 35216-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...