Ethyl-4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate 95%

Supplier: Ambeed
Warning

A163344-100MG A163344-250MG A163344-50MG
76783-962EA 69 USD
76783-962 76783-964 76783-966
Ethyl-4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate 95%
Ethyl-4-hydroxy-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₃
MDL Number: MFCD10699251
CAS Number: 187724-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR