2-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-((4,6-Dimethoxypyrimidin-2-yl)oxy)benzoic acid

A162980-1G A162980-250MG
76783-950EA 53.05 USD
76783-950 76783-952
2-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid 95%
2-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₅
MW: 276.25 g/mol
MDL Number: MFCD00203066
CAS Number: 110284-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR