Thieno[3,2-b]thiophene-2-carboxaldehyde 97%

Supplier: Ambeed
Warning

A162309-1G A162309-250MG A162309-5G
76783-914EA 88 USD
76783-914 76783-916 77600-302
Thieno[3,2-b]thiophene-2-carboxaldehyde 97%
Thieno[3,2-b]thiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₄OS₂
MDL Number: MFCD01859924
CAS Number: 31486-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR