7-Benzyloxyindole 98%

Supplier: Ambeed
Warning

A161740-25G A161740-5G A161740-10G
76783-884EA 157 USD
76783-884 76783-886 77871-810
7-Benzyloxyindole 98%
7-Benzyloxyindole

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO
MDL Number: MFCD00037974
CAS Number: 20289-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR