5-Bromo-2-methylsulfanylpyrimidin-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-(methylthio)pyrimidin-4(3H)-one

A161462-1000G A161462-100G A161462-10G A161462-1G A161462-250MG A161462-25G A161462-5G
76783-870EA 1844.91 USD
76783-870 76783-872 76783-874 76783-876 76783-878 76783-880 76783-882
5-Bromo-2-methylsulfanylpyrimidin-4-ol 98%
5-Bromo-2-methylsulfanylpyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₅BrN₂OS
MW: 221.08 g/mol
MDL Number: MFCD00223721
CAS Number: 81560-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR