3-Chloro-2-pyrazinemethanol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloropyrazin-2-yl)methanol

A161434-250MG A161434-1G
76783-868EA 38.2 USD
76783-868 77667-264
3-Chloro-2-pyrazinemethanol 98%
3-Chloro-2-pyrazinemethanol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD11977628
CAS Number: 89283-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR