(S)-N-Boc-4-bromophenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-p-bromo-Phe-OH, Boc-Phe(4-Br)-OH

A161042-100G A161042-10G A161042-1G A161042-250MG A161042-25G A161042-500G A161042-5G
76783-840EA 297.06 USD
76783-840 76783-842 76783-844 76783-846 76783-848 76783-850 76783-852
(S)-N-Boc-4-bromophenylalanine 97%
(S)-N-Boc-4-bromophenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
MW: 344.2 g/mol
MDL Number: MFCD00237571
CAS Number: 62129-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR