N-Methyl-5-bromo-4,7-diazaindole 98%

Supplier: Ambeed
Warning

A160971-1G A160971-250MG
76783-834EA 221.73 USD
76783-834 76783-836
N-Methyl-5-bromo-4,7-diazaindole 98%
N-Methyl-5-bromo-4,7-diazaindole

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
MDL Number: MFCD12964054
CAS Number: 1217090-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR