Boc-3-bromo-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-3-bromo-D-phenylalanine, (R)-3-(3-Bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

A160574-1G A160574-250MG A160574-5G A160574-25G
76783-814EA 13.8 USD
76783-814 76783-816 76783-818 76783-820 77871-806
Boc-3-bromo-D-phenylalanine 97%
Boc-3-bromo-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
MW: 344.205 g/mol
MDL Number: MFCD01317704
CAS Number: 261360-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR