5,6-Difluoroindoline 97%

Supplier: Ambeed
Warning

A160475-100MG A160475-1G A160475-250MG A160475-5G
76783-796EA 16.98 USD
76783-796 76783-798 76783-800 76783-802
5,6-Difluoroindoline 97%
5,6-Difluoroindoline

Precaución: for Research Use Only

Formula: C₈H₇F₂N
MW: 155.145 g/mol
MDL Number: MFCD01075221
CAS Number: 954255-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR