3-Aminooxetane hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Oxetan-3-amine hydrochloride

A159558-100MG A159558-250MG A159558-25G A159558-5G A159558-1G A159558-100G
76783-776EA 5 USD
76783-776 76783-778 76783-780 76783-782 77567-322 77871-798
3-Aminooxetane hydrochloride 95%
3-Aminooxetane hydrochloride

Precaución: For research use only.

Formula: C₃H₇NO·ClH
MW: 109.555 g/mol
MDL Number: MFCD09056831
CAS Number: 491588-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR