tert-Butyl-4-(5-aminopyridin-2-yl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Aminopyridin-2-yl)piperazine, N1-BOC protected, tert-Butyl 4-(5-aminopyridin-2-yl)piperazine-1-carboxylate

A159206-100MG A159206-1G A159206-250MG A159206-5G
76783-760EA 10.61 USD
76783-760 76783-762 76783-764 76783-766
tert-Butyl-4-(5-aminopyridin-2-yl)piperazine-1-carboxylate 95%
tert-Butyl-4-(5-aminopyridin-2-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₄O₂
MW: 278.3501 g/mol
MDL Number: MFCD05864841
CAS Number: 119285-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR