3-Bromo-4-chloro-7-azaindole 96%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine

A158827-100G A158827-10G A158827-1G A158827-250MG A158827-25G A158827-5G
76783-734EA 611 USD
76783-734 76783-736 76783-738 76783-740 76783-742 76783-744
3-Bromo-4-chloro-7-azaindole 96%
3-Bromo-4-chloro-7-azaindole

Precaución: for Research Use Only

Formula: C7H4BrClN2
MW: 231.4800 g/mol
MDL Number: MFCD09880132
CAS Number: 1000340-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR