5-Amino-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide 98%

Supplier: Ambeed
Warning

A158260-100G A158260-1G A158260-25G A158260-500G A158260-5G
76783-706EA 95 USD
76783-706 76783-708 76783-710 76783-712 76783-714
5-Amino-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide 98%
5-Amino-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈I₃N₃O₆
MDL Number: MFCD08063355
CAS Number: 76801-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR