Eltrombopag 99+%

Supplier: Ambeed
Warning

Synonyms: (Z)-3'-(2-(1-(3,4-Dimethylphenyl)-3-methyl-5-oxo-1H-pyrazol-4(5H)-ylidene)hydrazinyl)-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 3-[(5Z)-5-[[2-(3,4-dimethylphenyl)-5-methyl-3-oxo-1H-pyrazol-4-yl]hydrazinylidene]-6-oxocyclohexa-1,3-dien-1-yl]benzoic acid

A157281-100MG A157281-10MG A157281-1G A157281-1MG A157281-250MG A157281-25G A157281-50MG A157281-5G
76783-660EA 27.59 USD
76783-660 76783-662 76783-664 76783-666 76783-668 76783-670 76783-672 76783-674
Eltrombopag 99+%
Eltrombopag

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₄O₄
MW: 442.475 g/mol
MDL Number: MFCD20926253
CAS Number: 496775-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR