(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-D-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-D-aspartate, Fmoc-D-Asp(OtBu)-OH
A157057-100G
A157057-10G
A157057-1G
A157057-25G
A157057-500G
A157057-5G
76783-644EA
67
USD
76783-644
76783-646
76783-648
76783-650
76783-652
76783-654
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Precaución: for Research Use Only
Formula:
C₂₃H₂₅NO₆ |
MDL Number:
MFCD00077050 CAS Number: 112883-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...