ZSTK474 ≥98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(6-(2-(Difluoromethyl)-1H-benzo[d]imidazol-1-yl)-1,3,5-triazine-2,4-diyl)dimorpholine

A156873-10MG A156873-1MG A156873-50MG
76783-638EA 19.1 USD
76783-638 76783-640 76783-642
ZSTK474 ≥98%
ZSTK474

Precaución: for Research Use Only

Formula: C₁₉H₂₁F₂N₇O₂
MW: 417.4 g/mol
MDL Number: MFCD08705334
CAS Number: 475110-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR