6-Bromo-3,4-dihydro-2H-S,S-dioxothiochromen-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-bromothiochroman 1,1-dioxide hydrochloride

A156804-1MG A156804-250MG
76783-628EA 5.31 USD
76783-628 76783-630
6-Bromo-3,4-dihydro-2H-S,S-dioxothiochromen-4-amine hydrochloride 97%
6-Bromo-3,4-dihydro-2H-S,S-dioxothiochromen-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClNO₂S
MW: 312.62 g/mol
MDL Number: MFCD11616744
CAS Number: 1172986-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR