Vanillyl butyl ether 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Butoxymethyl)-2-methoxyphenol

A156385-100G A156385-10G A156385-25G A156385-500G A156385-5G
76783-600EA 85 USD
76783-600 76783-602 76783-604 77667-208 77667-210
Vanillyl butyl ether 95%
Vanillyl butyl ether

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₃
MDL Number: MFCD00238529
CAS Number: 82654-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR