2-Chloro-6-methylpyrimidine-4-carbonitrile 97%

Supplier: Ambeed
Warning

A156243-100MG A156243-1G A156243-250MG
76783-594EA 49.87 USD
76783-594 76783-596 76783-598
2-Chloro-6-methylpyrimidine-4-carbonitrile 97%
2-Chloro-6-methylpyrimidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.569 g/mol
MDL Number: MFCD09878518
CAS Number: 16858-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR