5-Methylpyrazin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-methylpyrazine

A156010-10G A156010-1G A156010-250MG
76783-576EA 354.35 USD
76783-576 76783-578 76783-580
5-Methylpyrazin-2(1H)-one 98%
5-Methylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C5H6N2O
MW: 110.1139 g/mol
MDL Number: MFCD01693937
CAS Number: 20721-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR