tert-Butyl (5-bromopyrazin-2-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromopyrazine, 2-BOC protected

A155925-10G A155925-250MG
76783-572EA 371 USD
76783-572 76783-574
tert-Butyl (5-bromopyrazin-2-yl)carbamate 98%
tert-Butyl (5-bromopyrazin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₂BrN₃O₂
MDL Number: MFCD08275683
CAS Number: 914349-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR