1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected 95%
Supplier: Ambeed
|
Warning
|
A155898-100MG
A155898-10G
A155898-1G
A155898-250MG
A155898-5G
76783-562EA
10
USD
76783-562
76783-564
76783-566
76783-568
76783-570
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected 95%
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
Precaución: for Research Use Only
Formula:
C₁₈H₂₅BN₂O₄ |
MDL Number:
MFCD08437629 CAS Number: 864771-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...